• USA Home
  • Search Results

  • phosphoenolpyruvicacidmonosodiumsalthydrate19002anhydrousbasis53823680anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phospho(enol)pyruvic acid monosodium salt hydrate

1 Products
Synonym:   2-(Phosphonooxy)-2-propenoic acid monosodium, Monosodium phosphoenolpyruvate, mono-Sodium phosphoenolpyruvate hydrate
Image of Phospho(enol)pyruvic acid monosodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C3H4NaO6P · xH2O
Molecular Weight:
 190.02 (anhydrous basis)
 Beilstein/REAXYS Number:
 4771585
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More