• USA Home
  • Search Results

  • phosphoricacid9800766438211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphoric acid

11 Products
Synonym:   Orthophosphoric acid
Image of Phosphoric acid
CAS Number:
Linear Formula:
 H3PO4
Molecular Weight:
 98.00
 Beilstein/REAXYS Number:
 1921286
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Condition
  Color
  Greener Alternative Principles
  Suitability
  Solubility
  Melting Point
  Density
  Agency/Method
  Application(s)
  Quality clarifier
  Purity Grade
  Product Line
  Concentration
  UV-Absorption Values
  Special Grade
  Physical Form
Show More