• USA Home
  • Search Results

  • phosphoruspentachloride208241002613811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphorus pentachloride

4 Products
Synonym:   Phosphorus(V) chloride
Image of Phosphorus pentachloride
CAS Number:
Linear Formula:
 PCl5
Molecular Weight:
 208.24
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  pH Value
  Vapor pressure
  Potency
  Melting Point
  Density
Show More