• USA Home
  • Search Results

  • phosphoruspentoxide14194131456311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphorus pentoxide

6 Products
Synonym:   Phosphoric anhydride, Phosphorus(V) oxide
Image of Phosphorus pentoxide
CAS Number:
Linear Formula:
 P2O5
Molecular Weight:
 141.94
 EC Number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Quality clarifier
  Assay
Show More