• USA Home
  • Search Results

  • phosphorusvoxychloride153331002587311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphorus(V) oxychloride

2 Products
Synonym:   Phosphorus(V) oxide chloride, Phosphoryl chloride
Image of Phosphorus(V) oxychloride
CAS Number:
Linear Formula:
 POCl3
Molecular Weight:
 153.33
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Special Grade
  Product Line
  pH Value
Show More