• USA Home
  • Search Results

  • physcion2842652161911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Physcion

3 Products
Synonym:   1,8-Dihydroxy-3-methoxy-6-methylanthraquinone, 6-O-Methylemodin, 6-Methoxychrysophanic acid, Emodin-3-methyl ether, Parietin, Rheochrysidin
Image of Physcion
CAS Number:
Empirical Formula (Hill Notation):
 C16H12O5
Molecular Weight:
 284.26
 Beilstein/REAXYS Number:
 6770499
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More