• USA Home
  • Search Results

  • phytagel123457101052111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phytagel

1 Products
Synonym:   Agar substitute gelling agent, Gellan Gum
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.72
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More