• USA Home
  • Search Results

  • phytol29653754149311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phytol

2 Products
Synonym:   3,7,11,15-Tetramethyl-2-hexadecen-1-ol
Image of Phytol
CAS Number:
Linear Formula:
 CH3[CH(CH3)(CH2)3]3C(CH3)=CHCH2OH
Molecular Weight:
 296.53
 Beilstein/REAXYS Number:
 1726098
 EC Number:
 FEMA Number:
 4196
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More