• USA Home
  • Search Results

  • picricacid229108889111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Picric acid

1 Products
Synonym:   2,4,6-Trinitrophenol, PA
Image of Picric acid
CAS Number:
Linear Formula:
 (O2N)3C6H2OH
Molecular Weight:
 229.10
 Beilstein/REAXYS Number:
 423400
 Colour Index Number:
 10305
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
Show More