• USA Home
  • Search Results

  • pilocarpinenitrate2712714872111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pilocarpine nitrate

5 Products
Synonym:   Pilocarpine nitrate salt
Image of Pilocarpine nitrate
CAS Number:
Empirical Formula (Hill Notation):
 C11H16N2O2 · HNO3
Molecular Weight:
 271.27
 Beilstein/REAXYS Number:
 4171406
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Manufacturer designation
Show More