• USA Home
  • Search Results

  • pilocarpinenitratesalt2712714872111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pilocarpine nitrate salt

1 Products
Image of Pilocarpine nitrate salt
CAS Number:
Empirical Formula (Hill Notation):
 C11H16N2O2 · HNO3
Molecular Weight:
 271.27
 Beilstein/REAXYS Number:
 4171406
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More