• USA Home
  • Search Results

  • piododphenylalanine291096256175511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Iodo-D-phenylalanine

1 Products
Synonym:   2-Amino-3-(4-iodophenyl)-propanoic acid
Image of <I>p</I>-Iodo-<SC>D</SC>-phenylalanine
CAS Number:
Linear Formula:
 C9H10NO2I
Molecular Weight:
 291.09
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More