• USA Home
  • Search Results

  • pipecolinicacid1291653575111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pipecolinic acid

1 Products
Synonym:   (±)-Piperidine-2-carboxylic acid, 2-Piperidinecarboxylic acid, Homoproline, Pipecolic acid free base
Image of Pipecolinic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H11NO2
Molecular Weight:
 129.16
 Beilstein/REAXYS Number:
 81095
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P45850 Pipecolinic acid, 98%,