• USA Home
  • Search Results

  • piperazine2carboxylicaciddihydrochloride20307302215911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piperazine-2-carboxylic acid dihydrochloride

1 Products
Synonym:   (±)-Piperazine-2-carboxylic acid dihydrochloride
Image of Piperazine-<WBR>2-<WBR>carboxylic acid dihydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C5H10N2O2 · 2HCl
Molecular Weight:
 203.07
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More