• USA Home
  • Search Results

  • piperazine861411085011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piperazine

4 Products
Synonym:   1,4-Diazacyclohexane, Diethylenediamine
Image of Piperazine
CAS Number:
Empirical Formula (Hill Notation):
 C4H10N2
Molecular Weight:
 86.14
 Beilstein/REAXYS Number:
 102555
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39160301
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  pH Value
  UV-Absorption Values
  Application(s)
  Product Line
  Physical Form
  Solubility
Show More