• USA Home
  • Search Results

  • piperidine851511089411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piperidine

9 Products
Synonym:   Hexahydropyridine
Image of Piperidine
CAS Number:
Empirical Formula (Hill Notation):
 C5H11N
Molecular Weight:
 85.15
 Beilstein/REAXYS Number:
 102438
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Document Links
  Allergen
  Organoleptic property
  UV-Absorption Values
  Application(s)
  Agency/Method
  Viscosity
  Potency
  Explosion limit
  Density
  Autoignition temp
  Refractive Index
  pH Value
  Transition temperature
  Purity Grade
  Melting Point
  Vapor pressure
  Special Grade
  Solubility
Show More