• USA Home
  • Search Results

  • piperine285349462211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piperine

8 Products
Synonym:   (E,E)-5-(3,4-Methylenedioxyphenyl)-2,4-pentadienoylpiperidide, 1-Piperoylpiperidine
Image of Piperine
CAS Number:
Empirical Formula (Hill Notation):
 C17H19NO3
Molecular Weight:
 285.34
 Beilstein/REAXYS Number:
 90741
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
  Anal. Standard Form
  Application(s)
  Melting Point
  Manufacturer designation
  Assay
  Physical Form
Show More