• USA Home
  • Search Results

  • piperonal1501312057011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piperonal

4 Products
Synonym:   1,3-Benzodioxole-5-carboxaldehyde, 3,4-(Methylenedioxy)benzaldehyde, Heliotropin
Image of Piperonal
CAS Number:
Empirical Formula (Hill Notation):
 C8H6O3
Molecular Weight:
 150.13
 Beilstein/REAXYS Number:
 131691
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Assay
  Agency/Method
Show More