• USA Home
  • Search Results

  • piperonylacetate1941832661411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piperonyl acetate

1 Products
Synonym:   heliotropyl acetate
Image of Piperonyl acetate
CAS Number:
Empirical Formula (Hill Notation):
 C10H10O4
Molecular Weight:
 194.18
 Council of Europe no.:
 2068
 EC Number:
 FEMA Number:
 2912
 Flavis number:
 9.220
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
Show More