• USA Home
  • Search Results

  • piperonylbutoxide338445103611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piperonylbutoxide

4 Products
Synonym:   2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether, 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether
Image of Piperonylbutoxide
CAS Number:
Empirical Formula (Hill Notation):
 C19H30O5
Molecular Weight:
 338.44
 Beilstein/REAXYS Number:
 288063
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Solubility
  Potency
  Assay
  Application(s)
Show More