• USA Home
  • Search Results

  • pipessodiumsalt324351001067011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

PIPES sodium salt

1 Products
Synonym:   1,4-Piperazinediethanesulfonic acid sodium salt, Piperazine-N,N′-bis(2-ethanesulfonic acid) sodium salt
Image of PIPES sodium salt
CAS Number:
Linear Formula:
 C8H17N2O6S2Na
Molecular Weight:
 324.35
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Solubility
Show More