• USA Home
  • Search Results

  • piroxicam331353632290411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Piroxicam

6 Products
Synonym:   4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2H-1,2-benzothiazine 1,1-dioxide
Image of Piroxicam
CAS Number:
Empirical Formula (Hill Notation):
 C15H13N3O4S
Molecular Weight:
 331.35
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More