• USA Home
  • Search Results

  • pk11195352868553275811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

PK 11195

1 Products
Synonym:   1-(2-Chlorophenyl)-N-methyl-N-(1-methylpropyl)-3-isoquinolinecarboxamide
Image of PK 11195
CAS Number:
Empirical Formula (Hill Notation):
 C21H21ClN2O
Molecular Weight:
 352.86
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
C0424 PK 11195,