• USA Home
  • Search Results

  • plasticadditive0412345801307811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Plastic additive 04

1 Products
Synonym:   ESBO, Epoxidized soya bean oil, Soybean oil, epoxidized
CAS Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More