• USA Home
  • Search Results

  • plasticadditive1077520170970211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Plastic additive 10

1 Products
Synonym:   1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene
Image of Plastic additive 10
CAS Number:
Linear Formula:
 (CH3)3C6[CH2C6H2[C(CH3)3]2OH]3
Molecular Weight:
 775.20
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More