• USA Home
  • Search Results

  • plasticadditive1795053250966311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Plastic additive 1

1 Products
Synonym:   Ethylene bis[3,3-bis[3-(1,1-dimethylethyl)-4-hydroxyphenyl]butanoate]
Image of Plastic additive 1
CAS Number:
Empirical Formula (Hill Notation):
 C50H66O8
Molecular Weight:
 795.05
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More