• USA Home
  • Search Results

  • plasticadditive2117763668319811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Plastic additive 2

2 Products
Synonym:   Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Pentaerythritol tetrakis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate)
Image of Plastic additive 2
CAS Number:
Linear Formula:
 [HOC6H2[C(CH3)3]2CH2CH2CO2CH2]4C
Molecular Weight:
 1177.63
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More