• USA Home
  • Search Results

  • plasticadditive453086208279311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Plastic additive 4

2 Products
Synonym:   Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate
Image of Plastic additive 4
CAS Number:
Linear Formula:
 [(CH3)3C]2C6H2(OH)CH2CH2CO2(CH2)17CH3
Molecular Weight:
 530.86
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More