• USA Home
  • Search Results

  • platinumiiacetylacetonate393291517057711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Platinum(II) acetylacetonate

2 Products
Synonym:   2,4-Pentanedione platinum(II) derivative, Pt(acac)2
Image of Platinum(II) acetylacetonate
CAS Number:
Linear Formula:
 Pt(C5H7O2)2
Molecular Weight:
 393.29
 Beilstein Registry Number:
 4136189
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More