• USA Home
  • Search Results

  • platinumrhodiumalloy123451110771411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Platinum-rhodium alloy

1 Products
Synonym:   Pt16Rh13
Image of Platinum-rhodium alloy
CAS Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More