• USA Home
  • Search Results

  • pleuromutilin3785012565511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pleuromutilin

1 Products
Synonym:   Drosophilin B, Mutilin 14-glycolate, NSC 121145
Image of Pleuromutilin
CAS Number:
Empirical Formula (Hill Notation):
 C22H34O5
Molecular Weight:
 378.50
 EC Number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
SML0285 Pleuromutilin, ≥95%,