• USA Home
  • Search Results

  • poly2vinylnaphthalene123452840656611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(2-vinylnaphthalene)

3 Products
Synonym:   2-Vinylnaphthalene homopolymer, 2-Vinylnaphthalene polymer, Poly(β-vinylnaphthalene)
Image of Poly(2-<WBR>vinylnaphthalene)
CAS Number:
Linear Formula:
 [CH2CH(C10H7)]n
 MDL number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Transition temperature
Show More