• USA Home
  • Search Results

  • poly3hexylthiophene25diyl1234515607498511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(3-hexylthiophene-2,5-diyl)

2 Products
Synonym:   P3HT
Image of Poly(3-<WBR>hexylthiophene-<WBR>2,5-<WBR>diyl)
CAS Number:
Linear Formula:
 (C10H14S)n
 MDL number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  OPV Device Performance
  Physical Form
  Special Grade
  Solubility
  Miscellaneous Specifications
  Assay
  Semiconductor properties
  Molecular weight
  Fluorescence Test
  Orbital energy
Show More