• USA Home
  • Search Results

  • polyacrylicacid12345900301411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(acrylic acid)

7 Products
Image of Poly(acrylic acid)
CAS Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Physical Form
  Cross-linking
  Viscosity
Show More