• USA Home
  • Search Results

  • polyacrylicacidsodiumsalt12345900304711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(acrylic acid sodium salt)

5 Products
Synonym:   Poly(sodium acrylate), Sodium polyacrylate
Image of Poly(acrylic acid sodium salt)
CAS Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  pH Value
  Density
  Shelf Life
  Anal. Standard Form
  Application(s)
  Physical Form
  Special Grade
Show More