• USA Home
  • Search Results

  • polyacrylicacidsolution12345900301411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(acrylic acid) solution

3 Products
Image of Poly(acrylic acid) solution
CAS Number:
Linear Formula:
 (C3H4O2)n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Molecular weight
  Density
Show More