• USA Home
  • Search Results

  • polyanilineemeraldinebase123452523330111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyaniline (emeraldine base)

6 Products
Synonym:   Emeraldine base polyaniline
Image of Polyaniline (emeraldine base)
CAS Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Absorption
Show More