• USA Home
  • Search Results

  • polybutylacrylatesolution12345900349011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(butyl acrylate) solution

2 Products
Synonym:   Acrylic acid butyl ester polymer, Butyl acrylate homopolymer, Butyl acrylate polymer, Poly(n -butyl acrylate)
Image of Poly(butyl acrylate) solution
CAS Number:
Linear Formula:
 (C7H12O2)n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Concentration
Show More