• USA Home
  • Search Results

  • polychloroprene12345901098411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polychloroprene

2 Products
Image of Polychloroprene
CAS Number:
Linear Formula:
 [-CH2CH=C(Cl)CH2-]n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Viscosity
  Physical Form
Show More