• USA Home
  • Search Results

  • polydimethylsiloxane123456314862911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(dimethylsiloxane)

2 Products
Synonym:   Dimethylpolysiloxane
Image of Poly(dimethylsiloxane)
CAS Number:
Linear Formula:
 [Si(CH3)2O]n
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Anal. Standard Form
  Physical Form
  Application(s)
  Special Grade
Show More