• USA Home
  • Search Results

  • polydimethylsiloxane12345901600611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polydimethylsiloxane

2 Products
Synonym:   Dimethylpolysiloxane, Polydimethylsiloxane
Image of Polydimethylsiloxane
CAS Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
Show More