• USA Home
  • Search Results

  • polydlysinehydrobromide123452796499411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly-D-lysine hydrobromide

9 Products
Synonym:   PDL
Image of Poly-<SC>D</SC>-lysine hydrobromide
CAS Number:
Linear Formula:
 D-Lys-(D-Lys)n-D-Lys · xHBr
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Physical Form
  Titertest
  Sterilization
  Solubility
  Product Line
  Package quantity
Show More