• USA Home
  • Search Results

  • polyethylene12345900288411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylene

10 Products
Image of Polyethylene
CAS Number:
Linear Formula:
 H(CH2CH2)nH
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Anal. Standard Form
  Shelf Life
  Viscosity
  Application(s)
  Particle Dimension
  Hardness
  Special Grade
  Melt index
  Melting Point
  Transition temperature
  Density
  Physical Form
Show More