• USA Home
  • Search Results

  • polyethyleneglycol123452532268311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol)

67 Products
Synonym:   PEG
Image of Poly(ethylene glycol)
CAS Number:
Linear Formula:
 H(OCH2CH2)nOH
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Sterilization
  Biological Source
  Restriction
  Refractive Index
  Agency/Method
  Viscosity
  Anal. Standard Form
  Shelf Life
  UV-Absorption Condition
  Application(s)
  Product Line
  pH Value
  Density
  Melting Point
  Functional group
  Solubility
  UV-Absorption Values
  Special Grade
  Physical Form
Show More

Product #
Description
89510 Poly(ethylene glycol), BioUltra, 8,000,
202398 Poly(ethylene glycol), average Mn 400, Green Alternative
P5413 Poly(ethylene glycol), for molecular biology, average mol wt 8,000,
81260 Poly(ethylene glycol), average Mn 6,000,
P3015 Poly(ethylene glycol), average mol wt 200,
P2139 Poly(ethylene glycol), average mol wt 8,000, powder,
95904 Poly(ethylene glycol), BioUltra, 4,000,
P4338 Poly(ethylene glycol), BioXtra, average mol wt 3,350, powder,
202444 Poly(ethylene glycol), average Mn 3,350, powder,
81268 Poly(ethylene glycol), BioUltra, for molecular biology, 8,000,
81300 Poly(ethylene glycol), average Mn 20,000,
81255 Poly(ethylene glycol), BioUltra, 6,000,
95172 Poly(ethylene glycol), BioUltra, 20,000,
81240 Poly(ethylene glycol), average Mn 4,000, platelets,
81310 Poly(ethylene glycol), 35,000,
81253 Poly(ethylene glycol), BioUltra, for molecular biology, 6,000,
81172 Poly(ethylene glycol), tested according to Ph Eur, 400,
202371 Poly(ethylene glycol), average Mn 300,
P3265 Poly(ethylene glycol), average mol wt 400,
91893 Poly(ethylene glycol), BioUltra, 400,
84797 Poly(ethylene glycol), BioUltra, 2,000,
88276 Poly(ethylene glycol), BioUltra, 3,350,
94646 Poly(ethylene glycol), BioUltra, 35,000,
81280 Poly(ethylene glycol), 10,000,
81188 Poly(ethylene glycol), BioUltra, 1,000,
92897 Poly(ethylene glycol), BioUltra, 10,000,
202401 Poly(ethylene glycol), average Mn 600, waxy solid (moist),
90878 Poly(ethylene glycol), BioUltra, 300,
81210 Poly(ethylene glycol), average Mw 1,500,
88440 Poly(ethylene glycol), BioUltra, 200,
P6667 Poly(ethylene glycol), average mol wt 10,000,
87333 Poly(ethylene glycol), BioUltra, 600,
P3515 Poly(ethylene glycol), average Mn 950-1,050,
81242 Poly(ethylene glycol), tested according to Ph Eur, 4,000,
P5402 Poly(ethylene glycol), average mol wt 1,450,
81290 Poly(ethylene glycol), analytical standard, for GPC, 2,000,
81292 Poly(ethylene glycol), analytical standard, for GPC, 4,000,
81285 Poly(ethylene glycol), 12,000,
309028 Poly(ethylene glycol), average Mn 10,000, flakes,
P2906 Poly(ethylene glycol), Hybri-Max, mol wt 3,000-3,700, waxy solid, BioReagent, suitable for hybridoma,
295906 Poly(ethylene glycol), average Mn 2,050, chips,
373001 Poly(ethylene glycol), average Mn 4,600,
81227 Poly(ethylene glycol), BioUltra, 3,000,
202436 Poly(ethylene glycol), average Mn 1,305-1,595, waxy solid,
202452 Poly(ethylene glycol), average Mv ~8,000, powder (crystalline),
81293 Poly(ethylene glycol), analytical standard, for GPC, 6,000,
03394 Poly(ethylene glycol), tested according to Ph Eur, 6,000,
81275 Poly(ethylene glycol), tested according to Ph Eur, 20,000,
81288 Poly(ethylene glycol), analytical standard, for GPC, 1,000,
81291 Poly(ethylene glycol), analytical standard, for GPC, 3,000,
86101 Poly(ethylene glycol), BioUltra, 1,500,
03395 Poly(ethylene glycol), tested according to Ph Eur, 1,000,
81162 Poly(ethylene glycol), tested according to Ph Eur, 300,
81189 Poly(ethylene glycol), BioUltra, for molecular biology, 1,000,
96699 Poly(ethylene glycol), analytical standard, for GPC, 40,000,
81296 Poly(ethylene glycol), analytical standard, for GPC, 12,000,
81294 Poly(ethylene glycol), analytical standard, for GPC, 8,000,
03557 Poly(ethylene glycol), tested according to Ph Eur, 35,000,
81298 Poly(ethylene glycol), analytical standard, for GPC, 20,000,
81289 Poly(ethylene glycol), analytical standard, for GPC, 1,500,
81284 Poly(ethylene glycol), analytical standard, for GPC, 400,
81201 Poly(ethylene glycol), analytical standard, for GPC, 108,000,
81286 Poly(ethylene glycol), analytical standard, for GPC, 600,
81206 Poly(ethylene glycol), analytical standard, for GPC, 218,000,
81283 Poly(ethylene glycol), analytical standard, for GPC, 300,
03473 Poly(ethylene glycol), analytical standard, for GPC, 40,000 certified according to DIN,
81282 Poly(ethylene glycol), analytical standard, for GPC, 200,