• USA Home
  • Search Results

  • polyethyleneglycol16acetylenedendrongeneration31234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol), 16 acetylene dendron, generation 3

2 Products
Synonym:   Bis-MPA poly(ethylene glycol), Bis[2,2-bis(hydroxymethyl)propanoic acid] PEG, Dendron functionalized PEG acetylene, generation 3, PEG-6000, PFD-G3-PEG6k-Acetylene
Image of Poly(ethylene glycol), 16 acetylene dendron, generation 3
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Functional group
  Miscellaneous Specifications
  ID clarifier
  Feature
Show More