• USA Home
  • Search Results

  • polyethyleneglycol20000123452532268311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylene glycol 20000

2 Products
Synonym:   PEG, Poly(ethylene glycol)
Image of Polyethylene glycol 20000
CAS Number:
Linear Formula:
 H(OCH2CH2)nOH
 EC Index Number:
 500-038-2
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  pH Value
  Vapor pressure
  Transition temperature
  Solubility
  Melting Point
  Density
  Bulk Density
  Autoignition temp
Show More