• USA Home
  • Search Results

  • polyethyleneglycol2mercaptoethyletheraceticacid1234516572981711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid

4 Products
Synonym:   mercaptoPEG acid, mercaptopoly(ethylene glycol) carboxylic acid, mercaptopolyoxyethylene acetic acid, thio-PEG, thiol-PEG-carboxylate, thiol-PEG-acid
Image of Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid
CAS Number:
Linear Formula:
 HSC2H4O(C2H4O)nCH2CO2H
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
  Transition temperature
  Polymer Architecture
  Molecular weight
  Functional group
Show More