• USA Home
  • Search Results

  • polyethyleneglycol3350123452532268311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylene glycol 3350

2 Products
Synonym:   PEG, Poly(ethylene glycol)
Image of Polyethylene glycol 3350
CAS Number:
Linear Formula:
 H(OCH2CH2)nOH
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More