• USA Home
  • Search Results

  • polyethyleneglycoldiacrylate123452657048911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol) diacrylate

8 Products
Synonym:   PEG diacrylate
Image of Poly(ethylene glycol) diacrylate
CAS Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
  Refractive Index
  Physical Form
  Density
  Transition temperature
  Reaction Suitability
  Polymer Architecture
  Molecular weight
  Additive
Show More