• USA Home
  • Search Results

  • polyethyleneglycolmethacrylate123452573686111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol) methacrylate

2 Products
Synonym:   Ethoxylated 2-hydroxyethyl methacrylate, PEGMA, Poly(ethylene oxide) monomethacrylate
Image of Poly(ethylene glycol) methacrylate
CAS Number:
Linear Formula:
 H2C=C(CH3)CO(OCH2CH2)nOH
 MDL number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
  Reaction Suitability
  Refractive Index
  Polymer Architecture
  Additive
  Molecular weight
  Density
Show More